missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. A18161G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18161G 1 g
A18165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18161G Supplier TCI America Supplier No. A18161G

Chemical Identifiers

CAS 106877-33-2
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.12
MDL Number MFCD01862656
InChI Key IBOZOWZSXZNIHI-UHFFFAOYSA-N
Synonym 5-amino-2-trifluoromethyl pyridine, 6-trifluoromethyl pyridin-3-amine, 3-amino-6-trifluoromethyl pyridine, 5-amino-2-trifluoromethylpyridine, 3-amino-6-trifluoromethylpyridine, 6-trifluoromethyl-3-pyridinamine, 3-pyridinamine, 6-trifluoromethyl, 6-trifluoromethyl-3-pyridylamine, 2-trifluoromethyl-5-aminopyridine, pubchem2997
PubChem CID 2737744
IUPAC Name 6-(trifluoromethyl)pyridin-3-amine
SMILES NC1=CC=C(N=C1)C(F)(F)F

Specifications

Melting Point 45°C
Quantity 1 g
Formula Weight 162.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-(trifluoromethyl)pyridine