missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-(trifluoromethyl)pyridine, 98%
Description
Employed in a convenient, one-pot, synthesis of azaindoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed in a convenient, one-pot, synthesis of azaindoles.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Employed in a convenient, one-pot, synthesis of azaindoles.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 39°C to 43°C |
| Flash Point | >110°C (230°F) |
| Quantity | 5 g |
| UN Number | UN2811 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 162.11 |
| Percent Purity | 98% |
| Chemical Name or Material | 5-Amino-2-(trifluoromethyl)pyridine |
Safety and Handling
| Hazard Category | H301-H315-H317-H319 |
| Hazard Statement | GHS H Statement H301-H315-H319-H317 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only