missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-(trifluoromethyl)pyridine, 98%

Catalog No. aah5696803
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AAH5696803 1 g
AAH5696806 5 g
2 options

Catalog No. AAH5696803

Supplier: Thermo Scientific Chemicals H5696803

Only null left

CAS: 106877-33-2 | C6H5F3N2 | 162.12 g/mol

Employed in a convenient, one-pot, synthesis of azaindoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed in a convenient, one-pot, synthesis of azaindoles.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 106877-33-2
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.12
MDL Number MFCD01862656
InChI Key IBOZOWZSXZNIHI-UHFFFAOYSA-N
Synonym 5-amino-2-trifluoromethyl pyridine, 6-trifluoromethyl pyridin-3-amine, 3-amino-6-trifluoromethyl pyridine, 5-amino-2-trifluoromethylpyridine, 3-amino-6-trifluoromethylpyridine, 6-trifluoromethyl-3-pyridinamine, 3-pyridinamine, 6-trifluoromethyl, 6-trifluoromethyl-3-pyridylamine, 2-trifluoromethyl-5-aminopyridine, pubchem2997
PubChem CID 2737744
IUPAC Name 6-(trifluoromethyl)pyridin-3-amine
SMILES NC1=CC=C(N=C1)C(F)(F)F

Specifications

Melting Point 39°C to 43°C
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 162.11
Percent Purity 98%
Chemical Name or Material 5-Amino-2-(trifluoromethyl)pyridine
Hazard Category H301-H315-H317-H319
Hazard Statement GHS H Statement
H301-H315-H319-H317
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only