missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-chloropyrimidine, 95%

Catalog No. AC434320050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434320050 5 g Glass bottle
AC434320010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434320050 Supplier Thermo Scientific Chemicals Supplier No. 434320050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56621-90-0 | C4H4ClN3 | 129.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56621-90-0
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.55
InChI Key DZBKIOJXVOECRA-UHFFFAOYSA-N
PubChem CID 12215993
IUPAC Name 2-chloropyrimidin-5-amine
SMILES C1=C(C=NC(=N1)Cl)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 160°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Synonym 5-amino-2-chloropyrimidine, 2-chloro-pyrimidin-5-ylamine, 2-chloro-5-aminopyrimidine, 5-pyrimidinamine, 2-chloro, 2-chloropyrimidine-5-ylamine, pubchem6963, acmc-1axfh, 2-chloropyrimidin-5-ylamine, 5-pyrimidinamine,2-chloro, 5-amino-2-chloro pyrimidine
Formula Weight 129.55
Percent Purity 95%
Physical Form Crystalline Powder or Solid
Chemical Name or Material 5-Amino-2-chloropyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Co

RUO – Research Use Only