missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-chlorobenzotrifluoride, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 103370250

Catalog No. AC103370250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

5-Amino-2-chlorobenzotrifluoride
320-51-4
100.0
134°C
98.5% min. (GC)
C7H5ClF3N
25g
4-chloro-3-trifluoromethyl aniline, 5-amino-2-chlorobenzotrifluoride, 3-trifluoromethyl-4-chloroaniline, 4-chloro-alpha,alpha,alpha-trifluoro-m-toluidine, 2-chloro-5-aminobenzotrifluoride, acbt, benzenamine, 4-chloro-3-trifluoromethyl, 2-chloro-5-aminebenzotrifluoride, 4-chloro-3-trifluoromethyl-phenylamine, 4-chloro-3-trifluoromethyl benzenamine
ASPDJZINBYYZRU-UHFFFAOYSA-N
4-chloro-3-(trifluoromethyl)aniline
67574
195.57
99%
98.5
85.0°C (3.0 mmHg)
Authentic
Glass bottle
H2NC6H3(Cl)CF3
MFCD00007846
Solubility in water: negligible
C1=CC(=C(C=C1N)C(F)(F)F)Cl
195.57
CHEBI:59245
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi

GHS Signal Word: Warning

EINECSNumber : 206-277-6

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions