missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-chloro-4-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog No. p-7053578
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6111514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6111514 Supplier Thermo Scientific Chemicals Supplier No. H6111514
Only null left

5-Amino-2-chloro-4-methylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-2-chloro-4-methylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 66909-38-4
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD03427661
InChI Key PLSMBLIGZJWLEJ-UHFFFAOYSA-N
Synonym 3-amino-6-chloro-4-picoline, 5-amino-2-chloro-4-methylpyridine, 5-amino-2-chloro-4-picoline, 3-amino-6-chloro-4-methyl-pyridine, 3-pyridinamine, 6-chloro-4-methyl, pubchem6206, 6-chloro-4-methyl-3-pyridinamine, 6-chloro-4-methyl-3-pyridylamine, 3-amino-6-chloro-4-methylpyridine, 6-chloro-4-methyl-3-pyridineamine
PubChem CID 2734425
IUPAC Name 6-chloro-4-methylpyridin-3-amine
SMILES CC1=CC(=NC=C1N)Cl

Specifications

Color Orange
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 142.59
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Amino-2-chloro-4-methylpyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only