missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-bromo-3-fluoropyridine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1256276-41-1
Molecular Formula C5H4BrFN2
Molecular Weight (g/mol) 191.003
MDL Number MFCD21099397
InChI Key DDJLNVAJTGIWNJ-UHFFFAOYSA-N
Synonym 5-amino-2-bromo-3-fluoropyridine, 3-pyridinamine, 6-bromo-5-fluoro
PubChem CID 71741382
IUPAC Name 6-bromo-5-fluoropyridin-3-amine
SMILES C1=C(C=NC(=C1F)Br)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH3315403
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH3315406
View Documents Promotion Details
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Commercially available 5-bromo-2-fluoropyridine was treated with LDA at -78°C to give the lithiated species. Fluoropyridine-based bromoacetamide reagent has been prepared and used in prosthetic group introduction for the labeling of oligonucleotides.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate(2-7°C ).
Specifications

Chemical Identifiers

1256276-41-1
191.003
DDJLNVAJTGIWNJ-UHFFFAOYSA-N
71741382
C1=C(C=NC(=C1F)Br)N
C5H4BrFN2
MFCD21099397
5-amino-2-bromo-3-fluoropyridine, 3-pyridinamine, 6-bromo-5-fluoro
6-bromo-5-fluoropyridin-3-amine

Specifications

1256276-41-1
MFCD21099397
DDJLNVAJTGIWNJ-UHFFFAOYSA-N
6-bromo-5-fluoropyridin-3-amine
71741382
97%
C5H4BrFN2
5-amino-2-bromo-3-fluoropyridine, 3-pyridinamine, 6-bromo-5-fluoro
C1=C(C=NC(=C1F)Br)N
191.003
191
5-Amino-2-bromo-3-fluoropyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only