Learn More
5-Amino-2-(4-morpholinyl)pyridine, 97%
CAS: 52023-68-4 | C9H13N3O | 179.223 g/mol
Supplier: Thermo Scientific Chemicals H5069906
Description
5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.
Notes
Store at room temperature. Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
Specifications
| 5-Amino-2-(4-morpholinyl)pyridine | |
| 127°C to 130°C | |
| MFCD00119562 | |
| 6-morpholinopyridin-3-amine, 3-amino-6-morpholinopyridine, 6-morpholino-3-pyridinamine, 6-morpholin-4-yl pyridin-3-amine, 6-morpholino pyridin-3-amine, 6-morpholinopyridine-3-amine, 3-pyridinamine, 6-4-morpholinyl, 6-morpholin-4-yl-3-pyridylamine, 5-amino-2-morpholin-4-ylpyridine, 5-amino-2-4-morpholinyl pyridine | |
| C1COCCN1C2=NC=C(C=C2)N | |
| 179.223 | |
| 179.22 |
| 52023-68-4 | |
| C9H13N3O | |
| 5 g | |
| VVTSPTCBHTWXMD-UHFFFAOYSA-N | |
| 6-morpholin-4-ylpyridin-3-amine | |
| 104059 | |
| 97% |
Chemical Identifiers
| 52023-68-4 | |
| 179.223 | |
| VVTSPTCBHTWXMD-UHFFFAOYSA-N | |
| 104059 | |
| C1COCCN1C2=NC=C(C=C2)N |
| C9H13N3O | |
| MFCD00119562 | |
| 6-morpholinopyridin-3-amine, 3-amino-6-morpholinopyridine, 6-morpholino-3-pyridinamine, 6-morpholin-4-yl pyridin-3-amine, 6-morpholino pyridin-3-amine, 6-morpholinopyridine-3-amine, 3-pyridinamine, 6-4-morpholinyl, 6-morpholin-4-yl-3-pyridylamine, 5-amino-2-morpholin-4-ylpyridine, 5-amino-2-4-morpholinyl pyridine | |
| 6-morpholin-4-ylpyridin-3-amine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
EINECSNumber : 257-613-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only