missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-(4-morpholinyl)pyridine, 97%

Catalog No. AAH50699MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50699MD 250 mg
AAH5069903 1 g
AAH5069906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH50699MD Supplier Thermo Scientific Chemicals Supplier No. H50699MD

CAS: 52023-68-4 | C9H13N3O | 179.223 g/mol

5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.

Notes
Store at room temperature. Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 52023-68-4
Molecular Formula C9H13N3O
Molecular Weight (g/mol) 179.223
MDL Number MFCD00119562
InChI Key VVTSPTCBHTWXMD-UHFFFAOYSA-N
Synonym 6-morpholinopyridin-3-amine, 3-amino-6-morpholinopyridine, 6-morpholino-3-pyridinamine, 6-morpholin-4-yl pyridin-3-amine, 6-morpholino pyridin-3-amine, 6-morpholinopyridine-3-amine, 3-pyridinamine, 6-4-morpholinyl, 6-morpholin-4-yl-3-pyridylamine, 5-amino-2-morpholin-4-ylpyridine, 5-amino-2-4-morpholinyl pyridine
PubChem CID 104059
IUPAC Name 6-morpholin-4-ylpyridin-3-amine
SMILES C1COCCN1C2=NC=C(C=C2)N

Specifications

Melting Point 127°C to 130°C
Quantity 250 mg
Formula Weight 179.22
Percent Purity 97%
Chemical Name or Material 5-Amino-2-(4-morpholinyl)pyridine
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 257-613-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only