missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-(4-aminophenyl)benzofuran 98.0+%, TCI America™
$406.37 - $1491.24
Chemical Identifiers
| CAS | 84102-58-9 |
|---|---|
| Molecular Formula | C14H12N2O |
| Molecular Weight (g/mol) | 224.26 |
| MDL Number | MFCD10000946 |
| InChI Key | FKMCNLLISWDXJQ-UHFFFAOYSA-N |
| Synonym | 2-(4-Aminophenyl)-5-benzofuranamine |
| PubChem CID | 11106990 |
| IUPAC Name | 2-(4-aminophenyl)-1-benzofuran-5-amine |
| SMILES | NC1=CC=C(C=C1)C1=CC2=CC(N)=CC=C2O1 |
Chemical Identifiers
| 84102-58-9 | |
| 224.26 | |
| FKMCNLLISWDXJQ-UHFFFAOYSA-N | |
| 11106990 | |
| NC1=CC=C(C=C1)C1=CC2=CC(N)=CC=C2O1 |
| C14H12N2O | |
| MFCD10000946 | |
| 2-(4-Aminophenyl)-5-benzofuranamine | |
| 2-(4-aminophenyl)-1-benzofuran-5-amine |
Specifications
| 84102-58-9 | |
| C14H12N2O | |
| 1 g | |
| FKMCNLLISWDXJQ-UHFFFAOYSA-N | |
| 2-(4-aminophenyl)-1-benzofuran-5-amine | |
| 11106990 | |
| ≥98.0% (GC) | |
| 5-Amino-2-(4-aminophenyl)benzofuran |
| 198°C | |
| MFCD10000946 | |
| 2-(4-Aminophenyl)-5-benzofuranamine | |
| NC1=CC=C(C=C1)C1=CC2=CC(N)=CC=C2O1 | |
| 224.26 | |
| 224.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No