missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-(4-aminophenyl)benzofuran 98.0+%, TCI America™

Catalog No. A19665G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19665G 5 g
A19661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19665G Supplier TCI America Supplier No. A19665G
Only null left

Chemical Identifiers

CAS 84102-58-9
Molecular Formula C14H12N2O
Molecular Weight (g/mol) 224.26
MDL Number MFCD10000946
InChI Key FKMCNLLISWDXJQ-UHFFFAOYSA-N
Synonym 2-(4-Aminophenyl)-5-benzofuranamine
PubChem CID 11106990
IUPAC Name 2-(4-aminophenyl)-1-benzofuran-5-amine
SMILES NC1=CC=C(C=C1)C1=CC2=CC(N)=CC=C2O1

Specifications

Melting Point 198°C
Quantity 5 g
Formula Weight 224.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-(4-aminophenyl)benzofuran