missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-(4-aminophenyl)benzofuran 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 198°C |
| Quantity | 1 g |
| Formula Weight | 224.26 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Amino-2-(4-aminophenyl)benzofuran |
Safety and Handling
| TSCA | No |