missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2,4,6-triiodoisophthalic Acid 96.0+%, TCI America™

Catalog No. A140625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A140625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A140625G Supplier TCI America Supplier No. A140625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35453-19-1
Molecular Formula C8H4I3NO4
Molecular Weight (g/mol) 558.836
MDL Number MFCD00190167
InChI Key JEZJSNULLBSYHV-UHFFFAOYSA-N
Synonym 5-amino-2,4,6-triiodoisophthalic acid, unii-a17g990rt0, 5-amino-2,4,6-triiodo-1,3-benzenedicarboxylic acid, 5-amino-2,4,6-iodoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino-2,4,6-triiodo, 5-amino-2,4,6-triiodoisophthalicacid, isophthalic acid, 5-amino-2,4,6-triiodo, acmc-209ih0, ksc224s6b, aronis25361
PubChem CID 3015783
IUPAC Name 5-amino-2,4,6-triiodobenzene-1,3-dicarboxylic acid
SMILES C1(=C(C(=C(C(=C1I)N)I)C(=O)O)I)C(=O)O

Specifications

Melting Point 270°C
Quantity 25 g
Formula Weight 558.84
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2,4,6-triiodoisophthalic Acid