missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2,4,6-triiodoisophthalic acid, 95%

Catalog No. p-7026263
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2217809 10 g
AAB2217818 50 g
AAB2217830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2217809 Supplier Thermo Scientific Chemicals Supplier No. B2217809
Only null left

CAS: 35453-19-1 | C8H4I3NO4 | 558.836 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35453-19-1
Molecular Formula C8H4I3NO4
Molecular Weight (g/mol) 558.836
MDL Number MFCD00190167
InChI Key JEZJSNULLBSYHV-UHFFFAOYSA-N
Synonym 5-amino-2,4,6-triiodoisophthalic acid, unii-a17g990rt0, 5-amino-2,4,6-triiodo-1,3-benzenedicarboxylic acid, 5-amino-2,4,6-iodoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino-2,4,6-triiodo, 5-amino-2,4,6-triiodoisophthalicacid, isophthalic acid, 5-amino-2,4,6-triiodo, acmc-209ih0, ksc224s6b, aronis25361
PubChem CID 3015783
IUPAC Name 5-amino-2,4,6-triiodobenzene-1,3-dicarboxylic acid
SMILES C1(=C(C(=C(C(=C1I)N)I)C(=O)O)I)C(=O)O

Specifications

Melting Point 265°C to 270°C
Quantity 10 g
Sensitivity Light and moisture sensitive
Formula Weight 558.84
Percent Purity 95%
Chemical Name or Material 5-Amino-2,4,6-triiodoisophthalic acid
Hazard Category H317-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 252-575-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only