missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2,2-difluoro-1,3-benzodioxole 98.0+%, TCI America™
$159.79 - $631.46
Chemical Identifiers
| CAS | 1544-85-0 |
|---|---|
| Molecular Formula | C7H5F2NO2 |
| Molecular Weight (g/mol) | 173.119 |
| MDL Number | MFCD00190144 |
| InChI Key | CVYQRDKVWVBOFP-UHFFFAOYSA-N |
| Synonym | 2,2-difluoro-5-aminobenzodioxole, 2,2-difluorobenzo d 1,3 dioxol-5-amine, 2,2-difluoro-5-amino-1,3-benzodioxole, 5-amino-2,2-difluorobenzodioxole, 5-amino-2,2-difluoro-1,3-benzodioxole, 2,2-difluoro-2h-1,3-benzodioxol-5-amine, 1,3-benzodioxol-5-amine, 2,2-difluoro, 3,4-difluoromethylenedioxy aniline, 3,4-difluoromethylene dioxy aniline, 2,2-difluoro-benzo 1,3 dioxol-5-ylamine |
| PubChem CID | 2736893 |
| IUPAC Name | 2,2-difluoro-1,3-benzodioxol-5-amine |
| SMILES | C1=CC2=C(C=C1N)OC(O2)(F)F |
Chemical Identifiers
| 1544-85-0 | |
| 173.119 | |
| CVYQRDKVWVBOFP-UHFFFAOYSA-N | |
| 2736893 | |
| C1=CC2=C(C=C1N)OC(O2)(F)F |
| C7H5F2NO2 | |
| MFCD00190144 | |
| 2,2-difluoro-5-aminobenzodioxole, 2,2-difluorobenzo d 1,3 dioxol-5-amine, 2,2-difluoro-5-amino-1,3-benzodioxole, 5-amino-2,2-difluorobenzodioxole, 5-amino-2,2-difluoro-1,3-benzodioxole, 2,2-difluoro-2h-1,3-benzodioxol-5-amine, 1,3-benzodioxol-5-amine, 2,2-difluoro, 3,4-difluoromethylenedioxy aniline, 3,4-difluoromethylene dioxy aniline, 2,2-difluoro-benzo 1,3 dioxol-5-ylamine | |
| 2,2-difluoro-1,3-benzodioxol-5-amine |
Specifications
| 1544-85-0 | |
| C7H5F2NO2 | |
| 1 g | |
| CVYQRDKVWVBOFP-UHFFFAOYSA-N | |
| 2,2-difluoro-1,3-benzodioxol-5-amine | |
| 2736893 | |
| ≥98.0% (GC,T) | |
| 5-Amino-2,2-difluoro-1,3-benzodioxole |
| 97°C | |
| MFCD00190144 | |
| 2,2-difluoro-5-aminobenzodioxole, 2,2-difluorobenzo d 1,3 dioxol-5-amine, 2,2-difluoro-5-amino-1,3-benzodioxole, 5-amino-2,2-difluorobenzodioxole, 5-amino-2,2-difluoro-1,3-benzodioxole, 2,2-difluoro-2h-1,3-benzodioxol-5-amine, 1,3-benzodioxol-5-amine, 2,2-difluoro, 3,4-difluoromethylenedioxy aniline, 3,4-difluoromethylene dioxy aniline, 2,2-difluoro-benzo 1,3 dioxol-5-ylamine | |
| C1=CC2=C(C=C1N)OC(O2)(F)F | |
| 173.119 | |
| 173.12 | |
| Liquid |
Safety and Handling
TSCA : No