missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2,2-difluoro-1,3-benzodioxole 98.0+%, TCI America™

Catalog No. A31075G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31075G 5 g
A31071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31075G Supplier TCI America Supplier No. A31075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1544-85-0
Molecular Formula C7H5F2NO2
Molecular Weight (g/mol) 173.119
MDL Number MFCD00190144
InChI Key CVYQRDKVWVBOFP-UHFFFAOYSA-N
Synonym 2,2-difluoro-5-aminobenzodioxole, 2,2-difluorobenzo d 1,3 dioxol-5-amine, 2,2-difluoro-5-amino-1,3-benzodioxole, 5-amino-2,2-difluorobenzodioxole, 5-amino-2,2-difluoro-1,3-benzodioxole, 2,2-difluoro-2h-1,3-benzodioxol-5-amine, 1,3-benzodioxol-5-amine, 2,2-difluoro, 3,4-difluoromethylenedioxy aniline, 3,4-difluoromethylene dioxy aniline, 2,2-difluoro-benzo 1,3 dioxol-5-ylamine
PubChem CID 2736893
IUPAC Name 2,2-difluoro-1,3-benzodioxol-5-amine
SMILES C1=CC2=C(C=C1N)OC(O2)(F)F

Specifications

Boiling Point 97°C
Quantity 5 g
Formula Weight 173.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Amino-2,2-difluoro-1,3-benzodioxole