missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1H-indazole, 98%

CAS: 19335-11-6 | C7H7N3 | 133.154 g/mol

$109.62 - $321.29

Chemical Identifiers

CAS 19335-11-6
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD00037975
InChI Key XBTOSRUBOXQWBO-UHFFFAOYSA-N
Synonym 5-aminoindazole, 5-amino-1h-indazole, 2h-indazol-5-amine, 5-indazolamine, 1h-indazole-5-amine, 1h-indazol-5-ylamine, indazol-5-ylamine, 1h-indazole, 5-amino, 5-aminobenzopyrazole, 5-amino-indazole
PubChem CID 88012
IUPAC Name 1H-indazol-5-amine
SMILES C1=CC2=C(C=C1N)C=NN2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0670506
View Documents
Thermo Scientific Chemicals
L0670506
5 g
Each for $109.62
Only null left
 
AAL0670514
View Documents
Thermo Scientific Chemicals
L0670514
25 g
Each for $321.29
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19335-11-6
133.154
XBTOSRUBOXQWBO-UHFFFAOYSA-N
88012
C1=CC2=C(C=C1N)C=NN2
C7H7N3
MFCD00037975
5-aminoindazole, 5-amino-1h-indazole, 2h-indazol-5-amine, 5-indazolamine, 1h-indazole-5-amine, 1h-indazol-5-ylamine, indazol-5-ylamine, 1h-indazole, 5-amino, 5-aminobenzopyrazole, 5-amino-indazole
1H-indazol-5-amine

Specifications

19335-11-6
Odorless
MFCD00037975
UN2811
5-aminoindazole, 5-amino-1h-indazole, 2h-indazol-5-amine, 5-indazolamine, 1h-indazole-5-amine, 1h-indazol-5-ylamine, indazol-5-ylamine, 1h-indazole, 5-amino, 5-aminobenzopyrazole, 5-amino-indazole
C1=CC2=C(C=C1N)C=NN2
133.154
133.15
5-Amino-1H-indazole
172°C to 178°C
C7H7N3
5 g
3259
XBTOSRUBOXQWBO-UHFFFAOYSA-N
1H-indazol-5-amine
88012
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 242-971-5

RTECSNumber : NK7711000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only