missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1H-indazole, 98%

Catalog No. AAL0670506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0670506 5 g
AAL0670514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0670506 Supplier Thermo Scientific Chemicals Supplier No. L0670506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19335-11-6 | C7H7N3 | 133.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19335-11-6
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD00037975
InChI Key XBTOSRUBOXQWBO-UHFFFAOYSA-N
Synonym 5-aminoindazole, 5-amino-1h-indazole, 2h-indazol-5-amine, 5-indazolamine, 1h-indazole-5-amine, 1h-indazol-5-ylamine, indazol-5-ylamine, 1h-indazole, 5-amino, 5-aminobenzopyrazole, 5-amino-indazole
PubChem CID 88012
IUPAC Name 1H-indazol-5-amine
SMILES C1=CC2=C(C=C1N)C=NN2

Specifications

Melting Point 172°C to 178°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 3259
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 5-Amino-1H-indazole
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 242-971-5
RTECSNumber NK7711000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only