missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-1-phenylpyrazole 98.0+%, TCI America™
$411.98 - $1291.79
Chemical Identifiers
| CAS | 826-85-7 |
|---|---|
| Molecular Formula | C9H9N3 |
| Molecular Weight (g/mol) | 159.192 |
| MDL Number | MFCD00020734 |
| InChI Key | ZVNYYNAAEVZNDW-UHFFFAOYSA-N |
| PubChem CID | 70006 |
| IUPAC Name | 2-phenylpyrazol-3-amine |
| SMILES | C1=CC=C(C=C1)N2C(=CC=N2)N |
Chemical Identifiers
| 826-85-7 | |
| 159.192 | |
| ZVNYYNAAEVZNDW-UHFFFAOYSA-N | |
| 2-phenylpyrazol-3-amine |
| C9H9N3 | |
| MFCD00020734 | |
| 70006 | |
| C1=CC=C(C=C1)N2C(=CC=N2)N |
Specifications
| 826-85-7 | |
| White-Yellow | |
| MFCD00020734 | |
| ZVNYYNAAEVZNDW-UHFFFAOYSA-N | |
| 2-phenylpyrazol-3-amine | |
| 70006 | |
| ≥98.0% (GC,T) | |
| 5-Amino-1-phenylpyrazole |
| 44°C | |
| C9H9N3 | |
| 5 g | |
| C1=CC=C(C=C1)N2C(=CC=N2)N | |
| 159.192 | |
| 159.19 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0086
RTECSNumber : UQ6108000
TSCA : No