missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-pentanol 95.0+%, TCI America™

Catalog No. p-7117101
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A087525G 25 g
A0875250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A087525G Supplier TCI America Supplier No. A087525G
Only null left

Chemical Identifiers

CAS 2508-29-4
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00008237
InChI Key LQGKDMHENBFVRC-UHFFFAOYSA-N
Synonym 5-amino-1-pentanol, 5-aminopentanol, 1-pentanol, 5-amino, pentanol, 5-amino, 5-amino-1-pentanol in water, 5-amino pentanol, 5-amino-l-pentanol, 5-aminopentanol-1, 5-hydroxypentylamine, 1-amino-5-pentanol
PubChem CID 75634
IUPAC Name 5-aminopentan-1-ol
SMILES C(CCN)CCO

Specifications

Color White-Yellow
Boiling Point 222°C
Quantity 25 g
UN Number 1759
Formula Weight 103.17
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Amino-1-pentanol