missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-1-naphthalenesulfonic Acid 98.0+%, TCI America™
Specifications
Specifications
| Color | Purple-Red |
| Quantity | 25 g |
| Formula Weight | 223.25 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Amino-1-naphthalenesulfonic Acid |
Safety and Handling
| EINECSNumber | (4)-0491 |
| RTECSNumber | QK1270500 |
| TSCA | Yes |