missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-methylbenzimidazole, 97%

Catalog No. p-7057109
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6646503 1 g
AAH6646506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6646503 Supplier Thermo Scientific Chemicals Supplier No. H6646503
Only null left

CAS: 10394-38-4 | C8H9N3 | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10394-38-4
Molecular Formula C8H9N3
Molecular Weight (g/mol) 147.18
MDL Number MFCD03164349
InChI Key IWBGBYZGEQUDBT-UHFFFAOYSA-N
Synonym 1-methylbenzoimidazol-5-amine, 1-methyl-1h-benzo d imidazol-5-amine, 1-methyl-1h-benzimidazol-5-amine, 1-methyl-1h-benzoimidazol-5-ylamine, 1-methyl-1h-1,3-benzodiazol-5-amine, 1-methyl-1,3-benzodiazol-5-amine, 1-methyl-1h-benzimidazol-5-ylamine, 1-methylbenzimidazole-5-ylamine, 1-methyl-5-aminobenzimidazole, 5-amino-1-methylbenzimidazole
PubChem CID 315499
IUPAC Name 1-methyl-1H-1,3-benzodiazol-5-amine
SMILES CN1C=NC2=CC(N)=CC=C12

Specifications

Quantity 1 g
Beilstein 128912
Formula Weight 147.08
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Amino-1-methylbenzimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only