missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB07485EA

 View more versions of this product

Catalog No. BTB07485EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile
96.5
97%
C10H7ClN4
10g
WIWSALMJHPGFDY-UHFFFAOYSA-N
5-amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile
639097
97%
51516-67-7
100.0
Amber Glass Bottle
MFCD00052031
5-amino-1-4-chlorophenyl-1h-pyrazole-4-carbonitrile, 5-amino-1-4-chlorophenyl pyrazole-4-carbonitrile, 1h-pyrazole-4-carbonitrile, 5-amino-1-4-chlorophenyl, maybridge1_001852, acmc-1b1ld, 5-amino-1-4-chloro-phenyl-1h-pyrazole-4-carbonit, 1-4-chlorophenyl-5-amino-1h-pyrazole-4-carbonitrile
NC1=C(C=NN1C1=CC=C(Cl)C=C1)C#N
218.64
218.65

Chemical Identifiers

51516-67-7
218.64
WIWSALMJHPGFDY-UHFFFAOYSA-N
639097
NC1=C(C=NN1C1=CC=C(Cl)C=C1)C#N
C10H7ClN4
MFCD00052031
5-amino-1-4-chlorophenyl-1h-pyrazole-4-carbonitrile, 5-amino-1-4-chlorophenyl pyrazole-4-carbonitrile, 1h-pyrazole-4-carbonitrile, 5-amino-1-4-chlorophenyl, maybridge1_001852, acmc-1b1ld, 5-amino-1-4-chloro-phenyl-1h-pyrazole-4-carbonit, 1-4-chlorophenyl-5-amino-1h-pyrazole-4-carbonitrile
5-amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions