missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1,3-diphenylpyrazole 98.0+%, TCI America™

Catalog No. A29041G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A29041G Supplier TCI America Supplier No. A29041G

Chemical Identifiers

CAS 5356-71-8
Molecular Formula C15H13N3
Molecular Weight (g/mol) 235.29
MDL Number MFCD00084878
InChI Key SXOFMEWDEKEVJU-UHFFFAOYSA-N
Synonym 1,3-diphenyl-1h-pyrazol-5-amine, 5-amino-1,3-diphenylpyrazole, 1h-pyrazol-5-amine, 1,3-diphenyl, pyrazole, 5-amino-1,3-diphenyl, 5-amino-1,3-diphenyl-1h-pyrazole, 1,3-diphenyl-1h-pyrazol-5-ylamine, 1,3-diphenylpyrazole-5-ylamine, maybridge1_004139, acmc-1apw6, cbmicro_003752
PubChem CID 199969
IUPAC Name 2,5-diphenylpyrazol-3-amine
SMILES C1=CC=C(C=C1)C2=NN(C(=C2)N)C3=CC=CC=C3

Specifications

Melting Point 130°C
Quantity 1 g
Formula Weight 235.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-1,3-diphenylpyrazole
RTECSNumber UQ5580000
TSCA No