missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1,3-diphenyl-1H-pyrazole, 97%

CAS: 5356-71-8 | C15H13N3 | 235.29 g/mol

$29.12 - $97.10

Chemical Identifiers

CAS 5356-71-8
Molecular Formula C15H13N3
Molecular Weight (g/mol) 235.29
MDL Number MFCD00084878
InChI Key SXOFMEWDEKEVJU-UHFFFAOYSA-N
Synonym 1,3-diphenyl-1h-pyrazol-5-amine, 5-amino-1,3-diphenylpyrazole, 1h-pyrazol-5-amine, 1,3-diphenyl, pyrazole, 5-amino-1,3-diphenyl, 5-amino-1,3-diphenyl-1h-pyrazole, 1,3-diphenyl-1h-pyrazol-5-ylamine, 1,3-diphenylpyrazole-5-ylamine, maybridge1_004139, acmc-1apw6, cbmicro_003752
PubChem CID 199969
IUPAC Name 2,5-diphenylpyrazol-3-amine
SMILES C1=CC=C(C=C1)C2=NN(C(=C2)N)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2046403
View Documents
Thermo Scientific Chemicals
B2046403
1 g
Each for $29.12
Only null left
 
AAB2046406
View Documents
Thermo Scientific Chemicals
B2046406
5 g
Each for $97.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5356-71-8
235.29
SXOFMEWDEKEVJU-UHFFFAOYSA-N
199969
C1=CC=C(C=C1)C2=NN(C(=C2)N)C3=CC=CC=C3
C15H13N3
MFCD00084878
1,3-diphenyl-1h-pyrazol-5-amine, 5-amino-1,3-diphenylpyrazole, 1h-pyrazol-5-amine, 1,3-diphenyl, pyrazole, 5-amino-1,3-diphenyl, 5-amino-1,3-diphenyl-1h-pyrazole, 1,3-diphenyl-1h-pyrazol-5-ylamine, 1,3-diphenylpyrazole-5-ylamine, maybridge1_004139, acmc-1apw6, cbmicro_003752
2,5-diphenylpyrazol-3-amine

Specifications

5356-71-8
Odorless
MFCD00084878
1,3-diphenyl-1h-pyrazol-5-amine, 5-amino-1,3-diphenylpyrazole, 1h-pyrazol-5-amine, 1,3-diphenyl, pyrazole, 5-amino-1,3-diphenyl, 5-amino-1,3-diphenyl-1h-pyrazole, 1,3-diphenyl-1h-pyrazol-5-ylamine, 1,3-diphenylpyrazole-5-ylamine, maybridge1_004139, acmc-1apw6, cbmicro_003752
C1=CC=C(C=C1)C2=NN(C(=C2)N)C3=CC=CC=C3
235.29
235.29
5-Amino-1,3-diphenyl-1H-pyrazole
129°C to 132°C
C15H13N3
1 g
SXOFMEWDEKEVJU-UHFFFAOYSA-N
2,5-diphenylpyrazol-3-amine
199969
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only