missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1,3-diphenyl-1H-pyrazole, 97%

Catalog No. AAB2046403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2046403 1 g
AAB2046406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2046403 Supplier Thermo Scientific Chemicals Supplier No. B2046403

CAS: 5356-71-8 | C15H13N3 | 235.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5356-71-8
Molecular Formula C15H13N3
Molecular Weight (g/mol) 235.29
MDL Number MFCD00084878
InChI Key SXOFMEWDEKEVJU-UHFFFAOYSA-N
Synonym 1,3-diphenyl-1h-pyrazol-5-amine, 5-amino-1,3-diphenylpyrazole, 1h-pyrazol-5-amine, 1,3-diphenyl, pyrazole, 5-amino-1,3-diphenyl, 5-amino-1,3-diphenyl-1h-pyrazole, 1,3-diphenyl-1h-pyrazol-5-ylamine, 1,3-diphenylpyrazole-5-ylamine, maybridge1_004139, acmc-1apw6, cbmicro_003752
PubChem CID 199969
IUPAC Name 2,5-diphenylpyrazol-3-amine
SMILES C1=CC=C(C=C1)C2=NN(C(=C2)N)C3=CC=CC=C3

Specifications

Melting Point 129°C to 132°C
Odor Odorless
Quantity 1 g
Formula Weight 235.29
Percent Purity 97%
Chemical Name or Material 5-Amino-1,3-diphenyl-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only