missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1,3-dimethyl-1H-pyrazole, 98%

Catalog No. p-7020876
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1298003 1 g
AAA1298006 5 g
AAA1298014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1298003 Supplier Thermo Scientific Chemicals Supplier No. A1298003

CAS: 3524-32-1 | C5H9N3 | 111.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3524-32-1
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.148
MDL Number MFCD00051651
InChI Key ZFDGMMZLXSFNFU-UHFFFAOYSA-N
Synonym 5-amino-1,3-dimethylpyrazole, 1,3-dimethyl-1h-pyrazol-5-amine, 1,3-dimethyl-1h-pyrazole-5yl-amine, 1h-pyrazol-5-amine, 1,3-dimethyl, 3-amino-2,5-dimethyl-2h-pyrazole, 2,5-dimethyl-2h-pyrazol-3-ylamine, 5-amino-1,3-dimethyl pyrazole, 1,3-dimethyl-1h-pyrazole-5-amine, 5-amino-1,3-dimethyl-1h-pyrazole, 1,3-dimethyl-5-aminopyrazole
PubChem CID 520721
IUPAC Name 2,5-dimethylpyrazol-3-amine
SMILES CC1=NN(C(=C1)N)C

Specifications

Melting Point 72°C to 77°C
Flash Point 88°C (190°F)
Quantity 1 g
Beilstein 113420
Sensitivity Hygroscopic
Formula Weight 111.14
Percent Purity 98%
Chemical Name or Material 5-Amino-1,3-dimethyl-1H-pyrazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only