missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%

Catalog No. p-7029784
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2728703 1 g
AAH2728706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2728703 Supplier Thermo Scientific Chemicals Supplier No. H2728703

CAS: 51516-68-8 | C10H7ClN4 | 218.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51516-68-8
Molecular Formula C10H7ClN4
Molecular Weight (g/mol) 218.64
MDL Number MFCD00128298
InChI Key VRKRSWGTAMOYEV-UHFFFAOYSA-N
Synonym 5-amino-1-3-chlorophenyl-1h-pyrazole-4-carbonitrile, 5-amino-1-3-chlorophenyl pyrazole-4-carbonitrile, 5-amino-1-3-chlorophenyl-1h-pyrazole-, 5-amino-1-3-chloro-phenyl-1h-pyrazole-4-carbonit, 1h-pyrazole-4-carbonitrile,5-amino-1-3-chlorophenyl, 1h-pyrazole-4-carbonitrile, 5-amino-1-3-chlorophenyl
PubChem CID 734496
IUPAC Name 5-amino-1-(3-chlorophenyl)pyrazole-4-carbonitrile
SMILES NC1=C(C=NN1C1=CC=CC(Cl)=C1)C#N

Specifications

Melting Point 183°C to 187°C
Quantity 1 g
Formula Weight 218.65
Percent Purity 97%
Chemical Name or Material 5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only