missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-1-(2-hydroxyethyl)pyrazole 99.0+%, TCI America™
$104.38 - $411.95
Chemical Identifiers
| CAS | 73616-27-0 |
|---|---|
| Molecular Formula | C5H9N3O |
| Molecular Weight (g/mol) | 127.147 |
| MDL Number | MFCD08063809 |
| InChI Key | IHQRJCVJAUKIEP-UHFFFAOYSA-N |
| Synonym | 5-amino-1-2-hydroxyethyl pyrazole, 2-5-aminopyrazol-1-yl ethanol, 5-amino-1-, a-hydroxyethyl pyrazole, 2-5-amino-1h-pyrazol-1-yl ethanol, 1h-pyrazole-1-ethanol, 5-amino, 2-5-amino-1h-pyrazol-1-yl ethan-1-ol, pubchem15051, ksc496i9l, 2-5-amino-1-pyrazolyl ethanol |
| PubChem CID | 320281 |
| IUPAC Name | 2-(5-aminopyrazol-1-yl)ethanol |
| SMILES | C1=C(N(N=C1)CCO)N |
Chemical Identifiers
| 73616-27-0 | |
| 127.147 | |
| IHQRJCVJAUKIEP-UHFFFAOYSA-N | |
| 320281 | |
| C1=C(N(N=C1)CCO)N |
| C5H9N3O | |
| MFCD08063809 | |
| 5-amino-1-2-hydroxyethyl pyrazole, 2-5-aminopyrazol-1-yl ethanol, 5-amino-1-, a-hydroxyethyl pyrazole, 2-5-amino-1h-pyrazol-1-yl ethanol, 1h-pyrazole-1-ethanol, 5-amino, 2-5-amino-1h-pyrazol-1-yl ethan-1-ol, pubchem15051, ksc496i9l, 2-5-amino-1-pyrazolyl ethanol | |
| 2-(5-aminopyrazol-1-yl)ethanol |
Specifications
| 73616-27-0 | |
| 140°C | |
| MFCD08063809 | |
| 5-amino-1-2-hydroxyethyl pyrazole, 2-5-aminopyrazol-1-yl ethanol, 5-amino-1-, a-hydroxyethyl pyrazole, 2-5-amino-1h-pyrazol-1-yl ethanol, 1h-pyrazole-1-ethanol, 5-amino, 2-5-amino-1h-pyrazol-1-yl ethan-1-ol, pubchem15051, ksc496i9l, 2-5-amino-1-pyrazolyl ethanol | |
| C1=C(N(N=C1)CCO)N | |
| 127.147 | |
| 127.15 | |
| Crystalline Powder |
| 108°C | |
| C5H9N3O | |
| 5 g | |
| IHQRJCVJAUKIEP-UHFFFAOYSA-N | |
| 2-(5-aminopyrazol-1-yl)ethanol | |
| 320281 | |
| ≥99.0% (GC) | |
| 5-Amino-1-(2-hydroxyethyl)pyrazole |
Safety and Handling
TSCA : No