New England Biolab Ltd NEB 5-alpha Competent E. coli (High Efficiency) 1 x 96 well plate (20 l/well)
Catalog No. NC0523896
NEB 5-alpha Competent E. coli (High Efficiency) is a derivative of the popular DH5. It is T1 phage resistant and endA deficient for high-quality plasmid preparations.
Catalog No.
NC0523896
Supplier
New England Biolab LtdSupplier No.
C2987P
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
NEB 5-alpha Competent E. coli (High Efficiency) is a derivative of the popular DH5. It is T1 phage resistant and endA deficient for high-quality plasmid preparations.