missing translation for 'onlineSavingsMsg'
Learn More

5-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate 96.0+%, TCI America™

Catalog No. A28951G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28951G 1 g
A28955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28951G Supplier TCI America Supplier No. A28951G
Only null left

Chemical Identifiers

CAS 133464-46-7
Molecular Formula C23H23NO6
Molecular Weight (g/mol) 409.44
MDL Number MFCD00190879
InChI Key LRBARFFNYOKIAX-UHFFFAOYNA-N
Synonym fmoc-glu oall-oh, fmoc-l-glutamic acid 5-allyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, ambotzfaa1372, 5-allyl n-fmoc-l-glutamate, n-fmoc-l-glutamic acid 5-allyl ester, fmoc-glu oall-oh hplc, 5-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 5-allyl ester, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid 5-allyl ester
PubChem CID 7019067
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid
SMILES OC(=O)C(CCC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 127°C
Color White
Quantity 1 g
Formula Weight 409.44
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate
TSCA No
Recommended Storage Refrigerator