Learn More
5-Acetylvaleric acid, 96%
CAS: 3128-07-2 | C7H12O3 | 144.17 g/mol
$143.15 - $884.42
Chemical Identifiers
| CAS | 3128-07-2 |
|---|---|
| Molecular Formula | C7H12O3 |
| Molecular Weight (g/mol) | 144.17 |
| MDL Number | MFCD00051479 |
| InChI Key | IZOQMUVIDMLRDC-UHFFFAOYSA-N |
| Synonym | 5-acetylvaleric acid, heptanoic acid, 6-oxo, 5-aceto valeric acid, 6-oxo-heptanoic acid, unii-4u902i8qzo, 5-acetylvalericacid, 6-oxoenanthic acid, 6-ketoheptanoic acid, 5-acetylpentanoic acid, # |
| PubChem CID | 98810 |
| IUPAC Name | 6-oxoheptanoic acid |
| SMILES | CC(=O)CCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1481006
|
Thermo Scientific Chemicals
A1481006 |
5 g |
Each for $143.15
|
|
|||||
|
AAA1481009
|
Thermo Scientific Chemicals
A1481009 |
10 g |
Each for $241.85
|
|
|||||
|
AAA1481018
|
Thermo Scientific Chemicals
A1481018 |
50 g |
Each for $884.42
|
|
|||||
Description
5-Acetylvaleric acid, is used as a reagent to synthesize new penicillins containing keto acids as side chains. It is also used to study the various metabolic pathways of 4-hydroxypentanoate and Levulinate. It is also used as an Pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Acetylvaleric acid, is used as a reagent to synthesize new penicillins containing keto acids as side chains. It is also used to study the various metabolic pathways of 4-hydroxypentanoate and Levulinate. It is also used as an Pharmaceutical intermediate.
Solubility
Soluble in water
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
Chemical Identifiers
| 3128-07-2 | |
| 144.17 | |
| IZOQMUVIDMLRDC-UHFFFAOYSA-N | |
| 98810 | |
| CC(=O)CCCCC(=O)O |
| C7H12O3 | |
| MFCD00051479 | |
| 5-acetylvaleric acid, heptanoic acid, 6-oxo, 5-aceto valeric acid, 6-oxo-heptanoic acid, unii-4u902i8qzo, 5-acetylvalericacid, 6-oxoenanthic acid, 6-ketoheptanoic acid, 5-acetylpentanoic acid, # | |
| 6-oxoheptanoic acid |
Specifications
| 3128-07-2 | |
| 138°C to 140°C (1.5 mmHg) | |
| 1.449 | |
| 5 g | |
| 1362931 | |
| 5-acetylvaleric acid, heptanoic acid, 6-oxo, 5-aceto valeric acid, 6-oxo-heptanoic acid, unii-4u902i8qzo, 5-acetylvalericacid, 6-oxoenanthic acid, 6-ketoheptanoic acid, 5-acetylpentanoic acid, # | |
| IZOQMUVIDMLRDC-UHFFFAOYSA-N | |
| 6-oxoheptanoic acid | |
| 98810 | |
| 96% |
| 34°C to 36°C | |
| C7H12O3 | |
| MFCD00051479 | |
| UN3261 | |
| Hygroscopic | |
| Soluble in water | |
| CC(=O)CCCCC(=O)O | |
| 144.17 | |
| 144.17 | |
| 5-Acetylvaleric acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 221-512-2
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only