missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylvaleric acid, 96%

Catalog No. AAA1481018
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1481018 50 g
AAA1481006 5 g
AAA1481009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1481018 Supplier Thermo Scientific Chemicals Supplier No. A1481018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3128-07-2 | C7H12O3 | 144.17 g/mol

5-Acetylvaleric acid, is used as a reagent to synthesize new penicillins containing keto acids as side chains. It is also used to study the various metabolic pathways of 4-hydroxypentanoate and Levulinate. It is also used as an Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Acetylvaleric acid, is used as a reagent to synthesize new penicillins containing keto acids as side chains. It is also used to study the various metabolic pathways of 4-hydroxypentanoate and Levulinate. It is also used as an Pharmaceutical intermediate.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.

Chemical Identifiers

CAS 3128-07-2
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00051479
InChI Key IZOQMUVIDMLRDC-UHFFFAOYSA-N
Synonym 5-acetylvaleric acid, heptanoic acid, 6-oxo, 5-aceto valeric acid, 6-oxo-heptanoic acid, unii-4u902i8qzo, 5-acetylvalericacid, 6-oxoenanthic acid, 6-ketoheptanoic acid, 5-acetylpentanoic acid, #
PubChem CID 98810
IUPAC Name 6-oxoheptanoic acid
SMILES CC(=O)CCCCC(=O)O

Specifications

Melting Point 34°C to 36°C
Boiling Point 138°C to 140°C (1.5 mmHg)
Refractive Index 1.449
Quantity 50 g
UN Number UN3261
Beilstein 1362931
Sensitivity Hygroscopic
Solubility Information Soluble in water
Formula Weight 144.17
Percent Purity 96%
Chemical Name or Material 5-Acetylvaleric acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 221-512-2
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only