missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylthiophene-2-carbonitrile, 97%

Catalog No. AAA1276502
Change view
Click to view available options
Quantity:
0.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1276502 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1276502 Supplier Thermo Scientific Chemicals Supplier No. A1276502
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 88653-55-8 | C7H5NOS | 151.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88653-55-8
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD00052314
InChI Key VSHPLUBHIUFLES-UHFFFAOYSA-N
Synonym 2-acetyl-5-cyanothiophene, 2-acetyl-5-cyanotiophene, 5-cyano-2-acetylthiophene, 5-acetyl-2-thiophenecarbonitrile, maybridge1_000101, acmc-20ao65, 2-cyano-5-acetyl-thiophene, 5-acetyl-thiophene-2-carbonitrile, 5-ethanoylthiophene-2-carbonitrile, 2-thiophenecarbonitrile, 5-acetyl-7ci,9ci
PubChem CID 2747565
IUPAC Name 5-acetylthiophene-2-carbonitrile
SMILES CC(=O)C1=CC=C(S1)C#N

Specifications

Melting Point 76°C to 80°C
Boiling Point 95°C to 97°C (0.5 mmHg)
Quantity 0.5 g
UN Number UN3439
Beilstein 1306961
Formula Weight 151.19
Percent Purity 97%
Chemical Name or Material 5-Acetylthiophene-2-carbonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only