missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-3-chloro-10,11-dihydrodibenzo[b,f]azepine 98.0+%, TCI America™

Catalog No. p-7117726
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
A20935G 5 g
A209325G 25 g
2 options

Catalog No. A20935G

Supplier: TCI America A20935G

Only null left

Chemical Identifiers

CAS 25961-11-9
Molecular Formula C16H14ClNO
Molecular Weight (g/mol) 271.744
MDL Number MFCD01632170
InChI Key NMZOSOMVILZBJL-UHFFFAOYSA-N
PubChem CID 117681
IUPAC Name 1-(2-chloro-5,6-dihydrobenzo[b][1]benzazepin-11-yl)ethanone
SMILES CC(=O)N1C2=CC=CC=C2CCC3=C1C=C(C=C3)Cl

Specifications

Melting Point 124°C
Color Yellow
Quantity 5 g
Synonym 5-Acetyl-3-chloro-2,2′C-iminodibenzyl
Formula Weight 271.74
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetyl-3-chloro-10,11-dihydrodibenzo[b,f]azepine