missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2-methoxypyridine 98.0+%, TCI America™

Catalog No. A27971G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A27971G Supplier TCI America Supplier No. A27971G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 213193-32-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD05664350
InChI Key RYOQZXOVBJIUSX-UHFFFAOYSA-N
Synonym 5-acetyl-2-methoxypyridine, 1-6-methoxypyridin-3-yl ethanone, 1-6-methoxypyridin-3-yl ethan-1-one, 1-6-methoxy-3-pyridyl ethanone, ethanone, 1-6-methoxy-3-pyridinyl, 3-acetyl-6-methoxypyridine, 1-6-methoxy-3-pyridinyl ethanone, pubchem16770, acmc-209fj4, 2-methoxy-5-acetylpyridine
PubChem CID 10534837
IUPAC Name 1-(6-methoxypyridin-3-yl)ethanone
SMILES CC(=O)C1=CN=C(C=C1)OC

Specifications

Melting Point 53°C
Color Yellow
Quantity 1 g
Formula Weight 151.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetyl-2-methoxypyridine