missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2-methoxypyridine, 97%

Catalog No. AC438170050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC438170050 5 g
AC438170010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438170050 Supplier Thermo Scientific Chemicals Supplier No. 438170050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 213193-32-9 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 213193-32-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
InChI Key RYOQZXOVBJIUSX-UHFFFAOYSA-N
Synonym 5-acetyl-2-methoxypyridine, 1-6-methoxypyridin-3-yl ethanone, 1-6-methoxypyridin-3-yl ethan-1-one, 1-6-methoxy-3-pyridyl ethanone, ethanone, 1-6-methoxy-3-pyridinyl, 3-acetyl-6-methoxypyridine, 1-6-methoxy-3-pyridinyl ethanone, pubchem16770, acmc-209fj4, 2-methoxy-5-acetylpyridine
PubChem CID 10534837
IUPAC Name 1-(6-methoxypyridin-3-yl)ethanone
SMILES CC(=O)C1=CN=C(C=C1)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 53.0°C to 54.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 5 g
Formula Weight 151.17
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 5-Acetyl-2-methoxypyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only