missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Acetyl-2-bromopyridine 98.0+%, TCI America™
$83.89 - $264.70
Chemical Identifiers
| CAS | 139042-59-4 |
|---|---|
| Molecular Formula | C7H6BrNO |
| Molecular Weight (g/mol) | 200.035 |
| MDL Number | MFCD04974527 |
| InChI Key | MUKKGHQBUKOMTD-UHFFFAOYSA-N |
| Synonym | 5-acetyl-2-bromopyridine, 1-6-bromopyridin-3-yl ethanone, 2-bromo-5-acetylpyridine, 1-6-bromo-pyridin-3-yl-ethanone, 1-6-bromopyridin-3-yl ethan-1-one, 3-acetyl-6-bromopyridine, ethanone, 1-6-bromo-3-pyridinyl, 1-6-bromopyrid-3-yl ethanone, 1-6-bromo-3-pyridyl ethanone, acmc-209cij |
| PubChem CID | 15668195 |
| IUPAC Name | 1-(6-bromopyridin-3-yl)ethanone |
| SMILES | CC(=O)C1=CN=C(C=C1)Br |
Chemical Identifiers
| 139042-59-4 | |
| 200.035 | |
| MUKKGHQBUKOMTD-UHFFFAOYSA-N | |
| 15668195 | |
| CC(=O)C1=CN=C(C=C1)Br |
| C7H6BrNO | |
| MFCD04974527 | |
| 5-acetyl-2-bromopyridine, 1-6-bromopyridin-3-yl ethanone, 2-bromo-5-acetylpyridine, 1-6-bromo-pyridin-3-yl-ethanone, 1-6-bromopyridin-3-yl ethan-1-one, 3-acetyl-6-bromopyridine, ethanone, 1-6-bromo-3-pyridinyl, 1-6-bromopyrid-3-yl ethanone, 1-6-bromo-3-pyridyl ethanone, acmc-209cij | |
| 1-(6-bromopyridin-3-yl)ethanone |
Specifications
| 139042-59-4 | |
| Yellow | |
| MFCD04974527 | |
| 5-acetyl-2-bromopyridine, 1-6-bromopyridin-3-yl ethanone, 2-bromo-5-acetylpyridine, 1-6-bromo-pyridin-3-yl-ethanone, 1-6-bromopyridin-3-yl ethan-1-one, 3-acetyl-6-bromopyridine, ethanone, 1-6-bromo-3-pyridinyl, 1-6-bromopyrid-3-yl ethanone, 1-6-bromo-3-pyridyl ethanone, acmc-209cij | |
| CC(=O)C1=CN=C(C=C1)Br | |
| 200.035 | |
| 200.04 | |
| Crystalline Powder |
| 126°C | |
| C7H6BrNO | |
| 1 g | |
| MUKKGHQBUKOMTD-UHFFFAOYSA-N | |
| 1-(6-bromopyridin-3-yl)ethanone | |
| 15668195 | |
| ≥98.0% (GC) | |
| 5-Acetyl-2-bromopyridine |
Safety and Handling
TSCA : No