missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2-amino-4-methylthiazole 98.0+%, TCI America™

Catalog No. p-7118127
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28871G 1 g
A28875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28871G Supplier TCI America Supplier No. A28871G
Only null left

Chemical Identifiers

CAS 30748-47-1
Molecular Formula C6H8N2OS
Molecular Weight (g/mol) 156.203
MDL Number MFCD00051952
InChI Key PKUKCASRNJIQNU-UHFFFAOYSA-N
Synonym 2-amino-4-methyl-5-acetylthiazole, 1-2-amino-4-methylthiazol-5-yl ethanone, 1-2-amino-4-methyl-1,3-thiazol-5-yl ethanone, 5-acetyl-2-amino-4-methylthiazole, 1-2-amino-4-methyl-1,3-thiazol-5-yl ethan-1-one, 1-2-amino-4-methyl-thiazol-5-yl-ethanone, 5-acetyl-2-amino-4-methyl-1,3-thiazole, ethanone, 1-2-amino-4-methyl-5-thiazolyl, 1-2-amino-4-methyl-1,3-thiazol-5-yl-1-ethanone, 5-acetyl-4-methyl-2-thiazolamine
PubChem CID 720882
IUPAC Name 1-(2-amino-4-methyl-1,3-thiazol-5-yl)ethanone
SMILES CC1=C(SC(=N1)N)C(=O)C

Specifications

Melting Point 266°C
Color White-Yellow
Quantity 1 g
Formula Weight 156.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetyl-2-amino-4-methylthiazole