missing translation for 'onlineSavingsMsg'
Learn More

5-Acetoxymethylfurfural 98.0+%, TCI America™

Catalog No. A14845G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14845G Supplier TCI America Supplier No. A14845G

Chemical Identifiers

CAS 10551-58-3
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00003233
InChI Key QAVITTVTXPZTSE-UHFFFAOYSA-N
Synonym 5-acetoxymethyl-2-furaldehyde, 5-formylfuran-2-yl methyl acetate, 5-acetoxymethylfurfural, 5-acetoxymethyl furfural, 5-formylfurfuryl acetate, 5-formyl-2-furyl methyl acetate, unii-5hh6180xyd, 5-acetoxymethyl furfural amf, 2-furancarboxaldehyde, 5-acetyloxy methyl
PubChem CID 66349
IUPAC Name (5-formylfuran-2-yl)methyl acetate
SMILES CC(=O)OCC1=CC=C(O1)C=O

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 122°C
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetoxymethylfurfural
EINECSNumber (5)-0044
TSCA No
Recommended Storage Freezer