missing translation for 'onlineSavingsMsg'
Learn More

5-Acetoxymethyl-2-furaldehyde, 97%

Catalog No. AC163400020
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC163400020 2 g
AC163400100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163400020 Supplier Thermo Scientific Chemicals Supplier No. 163400020

CAS: 10551-58-3 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10551-58-3
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00003233
InChI Key QAVITTVTXPZTSE-UHFFFAOYSA-N
Synonym 5-acetoxymethyl-2-furaldehyde, 5-formylfuran-2-yl methyl acetate, 5-acetoxymethylfurfural, 5-acetoxymethyl furfural, 5-formylfurfuryl acetate, 5-formyl-2-furyl methyl acetate, unii-5hh6180xyd, 5-acetoxymethyl furfural amf, 2-furancarboxaldehyde, 5-acetyloxy methyl
PubChem CID 66349
IUPAC Name (5-formylfuran-2-yl)methyl acetate
SMILES CC(=O)OCC1=CC=C(O1)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 52°C to 56°C
Color Brown
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 2 g
Beilstein 18, 15
Formula Weight 168.15
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material 5-Acetoxymethyl-2-furaldehyde, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
EINECSNumber 234-137-4

RUO – Research Use Only