missing translation for 'onlineSavingsMsg'
Learn More

5,8,11,14-Eicosatetraynoic acid

Catalog No. AAJ61624FPL
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61624FPL 20 mg
AAJ61624MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61624FPL Supplier Thermo Scientific Chemicals Supplier No. J61624FPL

A cyclooxygenase, lipoxygenase and cytochrome P450 inhibitor | CAS: 1191-85-1 | C20H24O2 | 296.41 g/mol

A cyclooxygenase, lipoxygenase and cytochrome P450 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1191-85-1
Molecular Formula C20H24O2
Molecular Weight (g/mol) 296.41
MDL Number MFCD00036967
InChI Key MGLDCXPLYOWQRP-UHFFFAOYSA-N
Synonym etya, 5,8,11,14-eicosatetraynoic acid, octadehydroarachidonic acid, eicosatetranoic acid, opera_id_402, spectrum5_001952, d0g9ip, bml2-f04, 5,8,11,14-eicosatetraynoicacid, 5,8,11,14-icosatetraynoic acid #
PubChem CID 1780
IUPAC Name icosa-5,8,11,14-tetraynoic acid
SMILES CCCCCC#CCC#CCC#CCC#CCCCC(=O)O

Specifications

Melting Point 81°C to 82°C
Color White
Linear Formula CH3(CH2)4(C≡CCH2)4CH2CH2CO2H
Quantity 20 mg
Beilstein 1798411
Formula Weight 296.4
Physical Form Solid
Chemical Name or Material 5,8,11,14-Eicosatetraynoic acid
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only