missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dimethoxycoumarin, 98%

Catalog No. AAB2322601
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2322601 0.25 g
AAB2322603 1 g
AAB2322606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2322601 Supplier Thermo Scientific Chemicals Supplier No. B2322601

CAS: 487-06-9 | C11H10O4 | 206.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 487-06-9
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.197
MDL Number MFCD00006870
InChI Key NXJCRELRQHZBQA-UHFFFAOYSA-N
Synonym 5,7-dimethoxycoumarin, citropten, limettin, citroptene, citraptene, limetin, 5,7-dimethoxy-2h-chromen-2-one, 5,7-dimethoxy-2-benzopyrone, 2h-1-benzopyran-2-one, 5,7-dimethoxy, coumarin, 5,7-dimethoxy
PubChem CID 2775
IUPAC Name 5,7-dimethoxychromen-2-one
SMILES COC1=CC(=C2C=CC(=O)OC2=C1)OC

Specifications

Melting Point 146°C to 149°C
Quantity 0.25 g
Merck Index 14,5492
Formula Weight 206.2
Percent Purity 98%
Chemical Name or Material 5,7-Dimethoxycoumarin
EINECSNumber 207-646-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only