missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichlorokynurenic acid

Catalog No. AAJ61138MA
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61138MA 10 mg
AAJ61138MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61138MA Supplier Thermo Scientific Chemicals Supplier No. J61138MA

CAS: 190908-40-8 | C10H7Cl2NO4 | 276.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 190908-40-8
Molecular Formula C10H7Cl2NO4
Molecular Weight (g/mol) 276.069
MDL Number MFCD11045951
InChI Key READYYZXDSXRQJ-UHFFFAOYSA-N
Synonym 5,7-dichlorokynurenic acid monohydrate, dcka hydrate, c10h5cl2no3 hydrate, 5,7-dichloro-4-hydroxyquinoline-2-carboxylic acid monohydrate, 5,7-dichlorokynurenic acid monohydrate, solid hplc, 2-quinolinecarboxylic acid, 5,7-dichloro-4-hydroxy-, monohydrate, 2-quinolinecarboxylicacid, 5,7-dichloro-4-hydroxy-, hydrate 1:1, 5,7-dichloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid-water 1/1
PubChem CID 23723057
IUPAC Name 5,7-dichloro-4-oxo-1H-quinoline-2-carboxylic acid;hydrate
SMILES C1=C(C=C2C(=C1Cl)C(=O)C=C(N2)C(=O)O)Cl.O

Specifications

Color Yellow
Quantity 10 mg
Solubility Information Soluble in dilute sodium hydroxide
Formula Weight 258.06
Physical Form Solid
Chemical Name or Material 5,7-Dichlorokynurenic acid monohydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 633-778-0
RTECSNumber VB1984600
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only