missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dibromo-8-hydroxyquinoline, 97%

Catalog No. p-7021801
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1428418 50 g
AAA1428430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1428418 Supplier Thermo Scientific Chemicals Supplier No. A1428418

CAS: 521-74-4 | C9H5Br2NO | 302.953 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 521-74-4
Molecular Formula C9H5Br2NO
Molecular Weight (g/mol) 302.953
MDL Number MFCD00006785
InChI Key ZDASUJMDVPTNTF-UHFFFAOYSA-N
Synonym broxyquinoline, 5,7-dibromo-8-hydroxyquinoline, 5,7-dibromo-8-quinolinol, broxyquinolin, dibromoxin, dibromoxine, dibromoxyquinoline, broxiquinoline, dibromoquin, broxykinolin
PubChem CID 2453
IUPAC Name 5,7-dibromoquinolin-8-ol
SMILES C1=CC2=C(C(=C(C=C2Br)Br)O)N=C1

Specifications

Melting Point 198°C to 203°C
Quantity 50 g
Beilstein 153624
Merck Index 14,1453
Formula Weight 302.96
Percent Purity 98%
Chemical Name or Material 5,7-Dibromo-8-hydroxyquinoline
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 208-317-8
RTECSNumber VC5300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only