missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dibromo-2,3-dihydrobenzofuran 97.0+%, TCI America™

Catalog No. D23365G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D23365G Supplier TCI America Supplier No. D23365G
Only null left

Chemical Identifiers

CAS 123266-59-1
Molecular Formula C8H6Br2O
Molecular Weight (g/mol) 277.943
MDL Number MFCD00191427
InChI Key NJQJUJHJJZMVNB-UHFFFAOYSA-N
Synonym 5,7-Dibromocoumaran
PubChem CID 2779135
IUPAC Name 5,7-dibromo-2,3-dihydro-1-benzofuran
SMILES C1COC2=C(C=C(C=C21)Br)Br

Specifications

Color Yellow
Quantity 5 g
Formula Weight 277.94
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5,7-Dibromo-2,3-dihydrobenzofuran