missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5(6)-TAMRA (5-(and-6)-Carboxytetramethylrhodamine), mixed isomers

Catalog No. c300
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C300 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C300 Supplier Invitrogen™ Supplier No. C300
Only null left

5(6)-TAMRA (5-(and-6)-Carboxytetramethylrhodamine), mixed isomers

5(6)-TAMRA contains a carboxylic acid that can be used to react with primary amines via carbodiimide activation of the carboxylic acid. This bright, orange-fluorescent dye produces conjugates with absorption/emission maxima of ∼555/580 nm.

Specifications

Quantity 100 mg
Product Type 5(6)-TAMRA
Content And Storage Store at room temperature and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Carboxylic Acid
Label or Dye TAMRA™ Isomers, TMR (Tetramethylrhodamine)
Label Type Classic Dyes
Emission 580

For Research Use Only. Not for use in diagnostic procedures.