missing translation for 'onlineSavingsMsg'
Learn More

5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine 98.0+%, TCI America™

Catalog No. d12011g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12011G Supplier TCI America Supplier No. D12011G
Only null left

Chemical Identifiers

CAS 1046-56-6
Molecular Formula C20H14N4
Molecular Weight (g/mol) 310.36
MDL Number MFCD00006462
InChI Key OTMYLOBWDNFTLO-UHFFFAOYSA-N
Synonym 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine, 5,6-diphenyl-3-2-pyridyl-1,2,4-triazine, 1,2,4-triazine, 5,6-diphenyl-3-2-pyridinyl, 5,6-diphenyl-3-pyridin-2-yl-1,2,4-triazine, as-triazine, 5,6-diphenyl-3-2-pyridyl, 5,6-diphenyl-3-2-pyridinyl-1,2,4-triazine, maybridge1_007490, bio7c7, acmc-1boy4, cbmicro_025706
PubChem CID 70588
IUPAC Name 5,6-diphenyl-3-(pyridin-2-yl)-1,2,4-triazine
SMILES C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=CC=C1)C1=CC=CC=N1

Specifications

Melting Point 190°C
Color Yellow
Quantity 1 g
Formula Weight 310.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine
RTECSNumber XY8598150
TSCA Yes