missing translation for 'onlineSavingsMsg'
Learn More

5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%

Catalog No. AAB2460103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2460103 1 g
AAB2460106 5 g
AAB2460114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2460103 Supplier Thermo Scientific Chemicals Supplier No. B2460103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1046-56-6 | C20H14N4 | 310.36 g/mol

5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 1046-56-6
Molecular Formula C20H14N4
Molecular Weight (g/mol) 310.36
MDL Number MFCD00006462
InChI Key OTMYLOBWDNFTLO-UHFFFAOYSA-N
Synonym 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine, 5,6-diphenyl-3-2-pyridyl-1,2,4-triazine, 1,2,4-triazine, 5,6-diphenyl-3-2-pyridinyl, 5,6-diphenyl-3-pyridin-2-yl-1,2,4-triazine, as-triazine, 5,6-diphenyl-3-2-pyridyl, 5,6-diphenyl-3-2-pyridinyl-1,2,4-triazine, maybridge1_007490, bio7c7, acmc-1boy4, cbmicro_025706
PubChem CID 70588
SMILES C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=CC=C1)C1=CC=CC=N1

Specifications

Melting Point 189°C to 192°C
Quantity 1 g
Beilstein 542151
Solubility Information Soluble in water.
IUPAC Name 5,6-diphenyl-3-pyridin-2-yl-1,2,4-triazine
Formula Weight 310.36
Percent Purity 99%
Chemical Name or Material 5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 213-878-7
RTECSNumber XY8598150
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only