missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Dimethyluracil 99.0+%, TCI America™
$126.76 - $423.14
Chemical Identifiers
| CAS | 26305-13-5 |
|---|---|
| Molecular Formula | C6H8N2O2 |
| Molecular Weight (g/mol) | 140.14 |
| MDL Number | MFCD00010708 |
| InChI Key | PZVLJGKJIMBYNP-UHFFFAOYSA-N |
| Synonym | 2,4-Dihydroxy-5,6-dimethylpyrimidine, 5,6-Dimethyl-2,4-pyrimidinediol |
| PubChem CID | 96031 |
| IUPAC Name | 5,6-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| SMILES | CC1=C(C)C(=O)NC(=O)N1 |
Chemical Identifiers
| 26305-13-5 | |
| 140.14 | |
| PZVLJGKJIMBYNP-UHFFFAOYSA-N | |
| 96031 | |
| CC1=C(C)C(=O)NC(=O)N1 |
| C6H8N2O2 | |
| MFCD00010708 | |
| 2,4-Dihydroxy-5,6-dimethylpyrimidine, 5,6-Dimethyl-2,4-pyrimidinediol | |
| 5,6-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione |
Specifications
| 26305-13-5 | |
| C6H8N2O2 | |
| 1 g | |
| PZVLJGKJIMBYNP-UHFFFAOYSA-N | |
| 5,6-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
| 96031 | |
| ≥99.0% (HPLC,T) | |
| 5,6-Dimethyluracil |
| White | |
| MFCD00010708 | |
| 2,4-Dihydroxy-5,6-dimethylpyrimidine, 5,6-Dimethyl-2,4-pyrimidinediol | |
| CC1=C(C)C(=O)NC(=O)N1 | |
| 140.14 | |
| 140.14 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : YQ9750000
TSCA : No